2,6-Pyridinedimethanol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,6-Bis(hydroxymethyl)pyridine

L02527-06 L02527-14
CAAAAL02527-06EA 101.12 CAD
CAAAAL02527-06 CAAAAL02527-14
2,6-Pyridinedimethanol 98%
2,6-Pyridinedimethanol

Purity: 98% CAS No.: 1195-59-1 Molecular Formula: C7H9NO2 Formula Weight: 139.15 Melting Point: 110-114° Boiling Point: 185°/15mm MDL No.: MFCD00006351

Formula: C₇H₉NO₂
MW: 139.15 g/mol
Boiling Pt: 185 °C (15 mmHg)
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00006351
CAS Number: 1195-59-1
EINECS: 214-803-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR