Diethyl allylmalonate 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L02286-14
L02286-22
CAAAAL02286-14EA
71.42
CAD
CAAAAL02286-14
CAAAAL02286-22
Diethyl allylmalonate 96%
Diethyl allylmalonate
Purity: 96%
CAS No.: 2049-80-1
Molecular Formula: C10H16O4
Formula Weight: 200.23
Flash Point: 92°(197°F)
Refractive Index: 1.4310
Boiling Point: 222-223°
Beilstein Reference: 1709466
MDL No.: MFCD00009155
Formula:
C₁₀H₁₆O₄ Boiling Pt: 222…223 °C Density: 1.016 Flash Pt: 92 °C(197 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009155 CAS Number: 2049-80-1 EINECS: 218-072-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...