7-Nitro-1-tetralone 97%
Supplier: Thermo Scientific Chemicals
L01853-04
L01853-09
CAAAAL01853-04EA
150.1
CAD
CAAAAL01853-04
CAAAAL01853-09
7-Nitro-1-tetralone 97%
7-Nitro-1-tetralone
Purity: 97%
CAS No.: 40353-34-2
Molecular Formula: C10H9NO3
Formula Weight: 191.19
Melting Point: 103-108°
Beilstein Reference: 1570515
Formula:
C₁₀H₉NO₃ Melting Pt: 103…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00019661 CAS Number: 40353-34-2 EINECS: 254-887-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...