3-Methyl-2-furoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L01763-03
L01763-06
CAAAAL01763-03EA
124.35
CAD
CAAAAL01763-03
CAAAAL01763-06
3-Methyl-2-furoic acid 98+%
3-Methyl-2-furoic acid
Purity: 98+%
CAS No.: 4412-96-8
Molecular Formula: C6H6O3
Formula Weight: 126.11
Melting Point: 133-137°
Beilstein Reference: 116183
Formula:
C₆H₆O₃ Melting Pt: 133…137 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014111 CAS Number: 4412-96-8 EINECS: 224-571-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...