3-Methyl-1H-pyrazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L01618-06
L01618-14
CAAAAL01618-06EA
55.93
CAD
CAAAAL01618-06
CAAAAL01618-14
3-Methyl-1H-pyrazole 97%
3-Methyl-1H-pyrazole
Purity: 97%
CAS No.: 1453-58-3
Molecular Formula: C4H6N2
Formula Weight: 82.11
Flash Point: 93°(199°F)
Refractive Index: 1.4960
Boiling Point: 203-205°
Beilstein Reference: 1454
MDL No.: MFCD00005240
Formula:
C₄H₆N₂ Boiling Pt: 203…205 °C Density: 1,020 Flash Pt: 93 °C(199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005240 CAS Number: 1453-58-3 EINECS: 215-925-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...