6-Amino-1,4-benzodioxane 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxin-6-amine, 1,4-Benzodioxan-6-amine

L01421-06 L01421-14 L01421-22
CAAAAL01421-06EA 61.78 CAD
CAAAAL01421-06 CAAAAL01421-14 CAAAAL01421-22
6-Amino-1,4-benzodioxane 99%
6-Amino-1,4-benzodioxane

Purity: 99% CAS No.: 22013-33-8 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Flash Point: 110°(230°F) Refractive Index: 1.5990 Melting Point: 26-30° Boiling Point: 116-118°/3mm Beilstein Reference: 6447 UN No.: 2811 MDL No.: MFCD00006824

Formula: C₈H₉NO₂
MW: 151.17 g/mol
Boiling Pt: 116…118 °C/3 mm
Melting Pt: 26…30 °C
Density: 1.231
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006824
CAS Number: 22013-33-8
EINECS: 244-718-4
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR