(1-Hexadecyl)tributylphosphonium bromide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Hexadecyltri-n-butylphosphonium bromide, Tributylhexadecylphosphonium bromide, Tributyl(hexadecyl)phosphonium bromide, (n-Hexadecyl)tri-n-butylphosphonium bromide
L01335-14
L01335-22
CAAAAL01335-06EA
101.83
CAD
CAAAAL01335-06
CAAAAL01335-14
CAAAAL01335-22
(1-Hexadecyl)tributylphosphonium bromide 98+%
(1-Hexadecyl)tributylphosphonium bromide
Fieser: 5,322 6,271
Purity: 98+%
CAS No.: 14937-45-2
Molecular Formula: C28H60BrP
Formula Weight: 507.67
Melting Point: 56-62°
Beilstein Reference: 4167404
Formula:
C₂₈H₆₀BrP MW: 507.65 g/mol Melting Pt: 56…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011775 CAS Number: 14937-45-2 EINECS: 239-014-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...