Isoquinoline-5-amine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Isoquinolin-5-amine, 5-Aminoisoquinoline

L01223-03 L01223-06 L01223-14
CAAAAL01223-03EA 64.46 CAD
CAAAAL01223-03 CAAAAL01223-06 CAAAAL01223-14
Isoquinoline-5-amine 99%
Isoquinoline-5-amine

Purity: 99% CAS No.: 1125-60-6 Molecular Formula: C9H8N2 Formula Weight: 144.18 Melting Point: 126-131° Beilstein Reference: 114465

Formula: C₉H₈N₂
Melting Pt: 126…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00006907
CAS Number: 1125-60-6
EINECS: 214-408-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR