m-Toluic hydrazide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L01043-06
L01043-14
CAAAAL01043-06EA
70.15
CAD
CAAAAL01043-06
CAAAAL01043-14
m-Toluic hydrazide 97%
m-Toluic hydrazide
Purity: 97%
CAS No.: 13050-47-0
Molecular Formula: C8H10N2O
Formula Weight: 150.18
Melting Point: 94-98°
Beilstein Reference: 2045125
Formula:
C₈H₁₀N₂O Melting Pt: 94…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014760 CAS Number: 13050-47-0 EINECS: 235-926-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...