Terephthalic acid dihydrazide 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Terephthalic Dihydrazide

L01017-06 L01017-14
CAAAAL01017-06EA 50.88 CAD
CAAAAL01017-06 CAAAAL01017-14
Terephthalic acid dihydrazide 95%
Terephthalic acid dihydrazide

Purity: 95% CAS No.: 136-64-1 Molecular Formula: C8H10N4O2 Formula Weight: 194.19 Melting Point: 300° Beilstein Reference: 982939

Formula: C₈H₁₀N₄O₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00014762
CAS Number: 136-64-1
EINECS: 205-253-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR