4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L00982-06
L00982-14
CAAAAL00982-06EA
146.96
CAD
CAAAAL00982-06
CAAAAL00982-14
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole 99+%
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole
Fieser: 4,19
Purity: 99+%
CAS No.: 1750-12-5
Molecular Formula: C2H6N6S
Formula Weight: 146.17
Melting Point: ca 220° dec.
Beilstein Reference: 134867
UN No.: 1325
MDL No.: MFCD00003098
Formula:
C₂H₆N₆S Melting Pt: ca 220 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003098 CAS Number: 1750-12-5 EINECS: 217-135-8 UN: 1325 ADR: "4.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...