o-Toluic hydrazide 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: o-Toluic acid hydrazide, 2-Methylbenzohydrazide

L00668-06 L00668-14
CAAAAL00668-06EA 111.07 CAD
CAAAAL00668-06 CAAAAL00668-14
o-Toluic hydrazide 98+%
o-Toluic hydrazide

Purity: 98+% CAS No.: 7658-80-2 Molecular Formula: C8H10N2O Formula Weight: 150.18 Melting Point: 122-124° Beilstein Reference: 1862809

Formula: C₈H₁₀N₂O
MW: 150.18
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00014756
CAS Number: 7658-80-2
EINECS: 231-623-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR