(4-Methoxybenzyl)triphenylphosphonium chloride 97%

Supplier: Thermo Scientific Chemicals
Warning

L00594-09 L00594-18
CAAAAL00594-09EA 116.45 CAD
CAAAAL00594-09 CAAAAL00594-18
(4-Methoxybenzyl)triphenylphosphonium chloride 97%
(4-Methoxybenzyl)triphenylphosphonium chloride

Purity: 97% CAS No.: 3462-97-3 Molecular Formula: C26H24ClOP Formula Weight: 418.91 Melting Point: 235-239° Beilstein Reference: 4173378

Formula: C₂₆H₂₄ClOP
MW: 418.91 g/mol
Melting Pt: 235…239 °C
Storage Temperature: Ambient
MDL Number: MFCD00031572
CAS Number: 3462-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR