1,8-Bis(dimethylamino)naphthalene 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N1,N1,N8,N8-Tetramethylnaphthalene-1,8-diamine

L00313-09 L00313-18
CAAAAL00313-09EA 67.15 CAD
CAAAAL00313-09 CAAAAL00313-18
1,8-Bis(dimethylamino)naphthalene 98+%
1,8-Bis(dimethylamino)naphthalene

Fieser: 3,22 4,35 6,50


Purity: 98+% CAS No.: 20734-58-1 Molecular Formula: C14H18N2 Formula Weight: 214.31 Flash Point: 110°(230°F) Melting Point: 47-51° Boiling Point: 144-145°/7mm Beilstein Reference: 396782 UN No.: 3263 MDL No.: MFCD00003920

Formula: C₁₄H₁₈N₂
MW: 214.31 g/mol
Boiling Pt: 144…145 °C/7 mm
Melting Pt: 47…51 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003920
CAS Number: 20734-58-1
EINECS: 244-001-6
UN: 3259
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR