Silver p-toluenesulfonate 98+%

Supplier: Thermo Scientific Chemicals
Warning

L00307-06 L00307-14
CAAAAL00307-06EA 96.06 CAD
CAAAAL00307-06 CAAAAL00307-14
Silver p-toluenesulfonate 98+%
Silver p-toluenesulphonate

Fieser: 1,1018 2,370 3,254 7,323 21,394


Purity: 98+% CAS No.: 16836-95-6 Molecular Formula: C7H7AgO3S Formula Weight: 279.07 Odor: Odorless UN No.: 3077 MDL No.: MFCD00007511

Formula: C₇H₇AgO₃S
Storage Temperature: Ambient
MDL Number: MFCD00007511
CAS Number: 16836-95-6
EINECS: 240-859-0
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR