Azidotrimethylsilane 94%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Trimethylsilyl azide
L00173-06
L00173-14
L00173-22
CAAAAL00173-06EA
66.68
CAD
CAAAAL00173-06
CAAAAL00173-14
CAAAAL00173-22
Azidotrimethylsilane 94%
Azidotrimethylsilane
Fieser: 1,1236 14,25 15,16 16,17 17,23 18,379 19,371 20,403 21,453
Purity: 94%
CAS No.: 4648-54-8
Molecular Formula: C3H9N3Si
Formula Weight: 115.21
Storage Temperature: 2°C to 8°C
Flash Point: 6°(42°F)
Refractive Index: 1.4140
Melting Point: -95°
Boiling Point: 95-99°
Beilstein Reference: 1903730
UN No.: 1992
MDL No.: MFCD00001986
Formula:
C₃H₉N₃Si MW: 115.21 g/mol Boiling Pt: 95…99 °C Melting Pt: –95 °C Density: 0.872 Flash Pt: 6 °C (42 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001986 CAS Number: 4648-54-8 EINECS: 225-078-5 UN: 1992 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...