3,5-Di-tert-butyl-o-benzoquinone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00169-06
L00169-14
CAAAAL00169-06EA
102.23
CAD
CAAAAL00169-06
CAAAAL00169-14
3,5-Di-tert-butyl-o-benzoquinone 98+%
3,5-Di-tert-butyl-o-benzoquinone
Fieser: 3,78 15,113 18,123
Purity: 98+%
CAS No.: 3383-21-9
Molecular Formula: C14H20O2
Formula Weight: 220.31
Melting Point: 113-117°
Beilstein Reference: 2047944
Formula:
C₁₄H₂₀O₂ Melting Pt: 113…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001647 CAS Number: 3383-21-9 EINECS: 222-189-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...