7-Bromo-1-tetralone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 7-Bromo-3,4-dihydronaphthalen-1(2H)-one

H50604-03 H50604-06
CAAAAH50604-03EA 191.18 CAD
CAAAAH50604-03 CAAAAH50604-06
7-Bromo-1-tetralone 97%
7-Bromo-1-tetralone

Purity: 97% CAS No.: 32281-97-3 Molecular Formula: C10H9BrO Formula Weight: 225.09 Melting Point: 73-77° MDL No.: MFCD02179287

Formula: C₁₀H₉BrO
MW: 225.08 g/mol
Melting Pt: 73…77 °C
Storage Temperature: Ambient
MDL Number: MFCD02179287
CAS Number: 32281-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR