1-(4-Bromophenyl)-2-pyrrolidinone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(4-Bromophenyl)pyrrolidin-2-one

H50563-03 H50563-06
CAAAAH50563-03EA 169.06 CAD
CAAAAH50563-03 CAAAAH50563-06
1-(4-Bromophenyl)-2-pyrrolidinone 98%
1-(4-Bromophenyl)-2-pyrrolidinone

Purity: 98% CAS No.: 7661-32-7 Molecular Formula: C10H10BrNO Formula Weight: 240.10 Boiling Point: 98-101° MDL No.: MFCD00138502

Formula: C₁₀H₁₀BrNO
MW: 240.10 g/mol
Boiling Pt: 98…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00138502
CAS Number: 7661-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR