4,6-Dichloro-2-methylpyrimidine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Methyl-4,6-dichloropyrimidine
H50493-03
H50493-06
CAAAAH50493-03EA
87.37
CAD
CAAAAH50493-03
CAAAAH50493-06
4,6-Dichloro-2-methylpyrimidine 97%
4,6-Dichloro-2-methylpyrimidine
Purity: 97%
CAS No.: 1780-26-3
Molecular Formula: C5H4Cl2N2
Formula Weight: 163.01
Melting Point: 41-45°
UN No.: 1759
MDL No.: MFCD00090472
Formula:
C₅H₄Cl₂N₂ Melting Pt: 41…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00090472 CAS Number: 1780-26-3 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...