5-Bromo-2-formylpyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

H50154-03 H50154-06
CAAAAH50154-03EA 99.38 CAD
CAAAAH50154-03 CAAAAH50154-06
5-Bromo-2-formylpyridine 99%
5-Bromo-2-formylpyridine

Purity: 99% CAS No.: 31181-90-5 Molecular Formula: C6H4BrNO Formula Weight: 186.01 Melting Point: 94-96° MDL No.: MFCD04112535

Formula: C₆H₄BrNO
Melting Pt: 94…96 °C
Storage Temperature: Ambient
MDL Number: MFCD04112535
CAS Number: 31181-90-5
UN: 2811
ADR: "6.1",III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR