2-Chloro-4-formylpyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H50098-03
H50098-06
CAAAAH50098-03EA
241.74
CAD
CAAAAH50098-03
CAAAAH50098-06
2-Chloro-4-formylpyridine 97%
2-Chloro-4-formylpyridine
Purity: 97%
CAS No.: 101066-61-9
Molecular Formula: C6H4ClNO
Formula Weight: 141.56
Melting Point: 46-49°
MDL No.: MFCD06651557
Formula:
C₆H₄ClNO Melting Pt: 46…49 °C Storage Temperature: Ambient |
MDL Number:
MFCD06651557 CAS Number: 101066-61-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...