3-Formyl-2-methoxypyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methoxynicotinaldehyde, 2-Methoxy-3-pyridinecarboxaldehyde, 2-Methoxy-pyridine-3-carbaldehyde, 2-Methoxypyridine-3-carboxaldehyde

H50087-03 H50087-06
CAAAAH50087-03EA 126.72 CAD
CAAAAH50087-03 CAAAAH50087-06
3-Formyl-2-methoxypyridine 97%
3-Formyl-2-methoxypyridine

Purity: 97% CAS No.: 71255-09-9 Molecular Formula: C7H7NO2 Formula Weight: 137.14 MDL No.: MFCD04115112

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115112
CAS Number: 71255-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR