1-Fluoro-4-ethynylbenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Ethynyl-4-fluorobenzene, (4-Fluorophenyl)acetylene, (4-Fluorophenyl)ethyne
H26269-03
H26269-06
CAAAH26269-03EA
87.06
CAD
CAAAH26269-03
CAAAH26269-06
1-Fluoro-4-ethynylbenzene 99%
1-Fluoro-4-ethynylbenzene
Purity: 99%
CAS No.: 766-98-3
Molecular Formula: C8H5F
Formula Weight: 120.12
Flash Point: 29°(85°F)
Refractive Index: 1.5170
Odor: Characteristic
Melting Point: 27-28°
Boiling Point: 55-56°/40mm
Beilstein Reference: 1099285
UN No.: 1325
MDL No.: MFCD00168823
Formula:
C₈H₅F MW: 120.13 g/mol Boiling Pt: 55…56 °C/40 mm Melting Pt: 27…28 °C Density: 1.048 Flash Pt: 29 °C (85 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00168823 CAS Number: 766-98-3 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...