5-Bromopicolinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromopyridine-2-carboxylic acid, 5-Bromo-2-pyridinecarboxylic acid

B25675-06 B25675-14
CAAAAB25675-03EA 58.03 CAD
CAAAAB25675-03 CAAAAB25675-06 CAAAAB25675-14
5-Bromopicolinic acid 98%
5-Bromopicolinic acid

Purity: 98% CAS No.: 30766-11-1 Molecular Formula: C6H4B2NO2 Formula Weight: 202.01 Melting Point: 173-175° MDL No.: MFCD00234149

Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Melting Pt: 173…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00234149
CAS Number: 30766-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR