4'-Aminobenzanilide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4′-Aminobenzanilide, 4-Aminobenzanilide

B25663-06 B25663-14
CAAAAB25663-06EA 103.81 CAD
CAAAAB25663-06 CAAAAB25663-14
4'-Aminobenzanilide 97%
4'-Aminobenzanilide

Purity: 97% CAS No.: 17625-83-1 Molecular Formula: C13H12N2O Formula Weight: 212.25 Melting Point: 127-131° MDL No.: MFCD00035777

Formula: C₁₃H₁₂N₂O
MW: 212.25 g/mol
Melting Pt: 127…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00035777
CAS Number: 17625-83-1
EINECS: 241-603-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR