Methyl-2-bromo-5-chlorobenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2-bromo-5-chlorobenzoate

B25309-14 B25309-22
CAAAAB25309-14EA 151.68 CAD
CAAAAB25309-14 CAAAAB25309-22
Methyl-2-bromo-5-chlorobenzoate 98%
Methyl 2-bromo-5-chlorobenzoate

Purity: 98% CAS No.: 27007-53-0 Molecular Formula: C8H6BrClO2 Formula Weight: 249.49 Melting Point: 37-40° MDL No.: MFCD00144763

Formula: C₈H₆BrClO₂
Melting Pt: 37…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00144763
CAS Number: 27007-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR