3,3-Dimethylglutaric anhydride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25286-06
B25286-14
CAAAAB25286-06EA
74.58
CAD
CAAAAB25286-06
CAAAAB25286-14
CAAAAB25286-22
3,3-Dimethylglutaric anhydride 97%
3,3-Dimethylglutaric anhydride
Purity: 97%
CAS No.: 4160-82-1
Molecular Formula: C7H10O3
Formula Weight: 142.15
Melting Point: 122-126°
Boiling Point: 181°/25mm
MDL No.: MFCD00006684
Formula:
C₇H₁₀O₃ Boiling Pt: 181 °C/25 mm Melting Pt: 122…126 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006684 CAS Number: 4160-82-1 EINECS: 223-998-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...