6,7-Dimethoxy-1-tetralone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25226-03
B25226-06
B25226-14
CAAAAB25226-03EA
130.98
CAD
CAAAAB25226-03
CAAAAB25226-06
CAAAAB25226-14
6,7-Dimethoxy-1-tetralone 97%
6,7-Dimethoxy-1-tetralone
Purity: 97%
CAS No.: 13575-75-2
Molecular Formula: C12H14O3
Formula Weight: 206.24
Melting Point: 98-100°
MDL No.: MFCD00134100
Formula:
C₁₂H₁₄O₃ Melting Pt: 98…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134100 CAS Number: 13575-75-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...