3-Methyl-3-oxetanemethanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methyl-3-oxethanemethanol

B25111-14 B25111-22
CAAAAB25111-14EA 122.45 CAD
CAAAAB25111-14 CAAAAB25111-22
3-Methyl-3-oxetanemethanol 97%
3-Methyl-3-oxetanemethanol

Fieser: 12,323


Purity: 97% CAS No.: 3143-02-0 Molecular Formula: C5H10O2 Formula Weight: 102.13 Storage Temperature: 2°C to 8°C Flash Point: 98°(208°F) Refractive Index: 1.4460 Boiling Point: 79-80°/40mm Beilstein Reference: 102773 MDL No.: MFCD00010273

Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 79…80 °C/40 mm
Density: 1.024
Flash Pt: 98 °C (208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010273
CAS Number: 3143-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR