2-Methyl-3-furoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24928-06
B24928-14
CAAAAB24928-06EA
94.48
CAD
CAAAAB24928-06
CAAAAB24928-14
CAAAAB24928-22
2-Methyl-3-furoic acid 98%
2-Methyl-3-furoic acid
Purity: 98%
CAS No.: 6947-94-0
Molecular Formula: C6H6O3
Formula Weight: 126.11
Melting Point: 97-99°
Beilstein Reference: 112229
Formula:
C₆H₆O₃ Melting Pt: 97…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00092314 CAS Number: 6947-94-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...