Methyl-2,6-dichlorobenzoate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2,6-dichlorobenzoate

B24912-06 B24912-03
AAB24912-06EA 200.66 CAD
AAB24912-06 AAB24912-03
Methyl-2,6-dichlorobenzoate 97%
Methyl 2,6-dichlorobenzoate
Formula: C₈H₆Cl₂O₂
MW: 205.04
Boiling Pt: 250 °C
Melting Pt: 25…30 °C
Storage Temperature: Ambient
MDL Number: MFCD01313701
CAS Number: 14920-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR