Diethyl phenylphosphonite 98%

Supplier: Thermo Scientific Chemicals
Warning

B24836-06 B24836-14
CAAAAB24836-06EA 224.36 CAD
CAAAAB24836-06 CAAAAB24836-14 CAAAAB24836-22
Diethyl phenylphosphonite 98%
Diethyl phenylphosphonite

Purity: 98% CAS No.: 1638-86-4 Molecular Formula: (CH3CH2O)2P(C6H5) Formula Weight: 198.20 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5100 Boiling Point: 64°/0.7mm Beilstein Reference: 2804900 MDL No.: MFCD00009086

Formula: C₁₀H₁₅O₂P
MW: 198.2 g/mol
Boiling Pt: 64 °C (0.7 mmHg)
Density: 1.032
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00009086
CAS Number: 1638-86-4
EINECS: 216-676-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR