Cyclohexyl phenyl ketone 98%

Supplier: Thermo Scientific Chemicals
Warning

B24816-06 B24816-14 B24816-22
CAAAAB24816-06EA 47.56 CAD
CAAAAB24816-06 CAAAAB24816-14 CAAAAB24816-22
Cyclohexyl phenyl ketone 98%
Cyclohexyl phenyl ketone

Purity: 98% CAS No.: 712-50-5 Molecular Formula: C13H16O Formula Weight: 188.27 Flash Point: 110°(230°F) Melting Point: 52-57° Boiling Point: 165-168°/20mm Beilstein Reference: 2046712 MDL No.: MFCD00001467

Formula: C₁₃H₁₆O
MW: 188.27 g/mol
Boiling Pt: 165…168 °C/20 mm
Melting Pt: 52…57 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001467
CAS Number: 712-50-5
EINECS: 211-923-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR