4-Ethynyltoluene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Ethynyl-4-methylbenzene, p-Tolylacetylene

B24812-06 B24812-14
CAAAAB24812-06EA 98.43 CAD
CAAAAB24812-06 CAAAAB24812-14
4-Ethynyltoluene 98%
4-Ethynyltoluene

Purity: 98% CAS No.: 766-97-2 Molecular Formula: C9H8 Formula Weight: 116.16 Flash Point: 49°(120°F) Refractive Index: 1.5470 Odor: Pungent Melting Point: ca 21° Boiling Point: 168-170° Beilstein Reference: 969653 UN No.: 3295 MDL No.: MFCD00008571

Formula: C₉H₈
Boiling Pt: 168…170 °C
Melting Pt: ca 21 °C
Density: 0.916
Flash Pt: 49 °C(120 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008571
CAS Number: 766-97-2
UN: 3295
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR