2-Amino-4-hydroxy-6-methylpyrimidine 98%
Supplier: Thermo Scientific Chemicals
B24604-18
B24604-30
CAAAB24604-18EA
98.59
CAD
CAAAB24604-18
CAAAB24604-30
2-Amino-4-hydroxy-6-methylpyrimidine 98%
2-Amino-4-hydroxy-6-methylpyrimidine
Purity: 98%
CAS No.: 3977-29-5
Molecular Formula: C5H7N3O
Formula Weight: 125.13
Melting Point: ca 305° dec.
Beilstein Reference: 3531
Formula:
C₅H₇N₃O Melting Pt: ca 305 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006095 CAS Number: 3977-29-5 EINECS: 223-612-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...