2,6-Dimethylpiperidin (mixture of cis- and trans-isomers) predominantly cis 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,6-Lupetidine

B24524-06 B24524-18
CAAAAB24524-06EA 31.44 CAD
CAAAAB24524-06 CAAAB24524-18
2,6-Dimethylpiperidin (mixture of cis- and trans-isomers) predominantly cis 99%
2,6-Dimethylpiperidin (mixture of cis- and trans-isomers)

Used as a base for the isolation of naturally-occurring carboxylic acids as their crystalline salts


Purity: 99% CAS No.: 504-03-0 Molecular Formula: C7H15N Formula Weight: 113.20 Flash Point: 11°(51°F) Refractive Index: 1.4400 Melting Point: -20° Boiling Point: 126-129° Beilstein Reference: 79827 UN No.: 2733 MDL No.: MFCD00066304

Formula: C₇H₁₅N
MW: 113.2 g/mol
Boiling Pt: 126…129 °C
Melting Pt: < –20 °C
Density: 0.832
Flash Pt: 11 °C(51 °F)
Storage Temperature: Ambient
MDL Number: MFCD00066304
CAS Number: 504-03-0
EINECS: 207-981-6
UN: 2733
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR