2-Aminoterephthalic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Aminoterephthalic Acid

B24441-06 B24441-14 B24441-22
CAAAB24441-06EA 48.51 CAD
CAAAB24441-06 CAAAB24441-14 CAAAB24441-22
2-Aminoterephthalic acid 99%
2-Aminoterephthalic acid

Purity: 99% CAS No.: 10312-55-7 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Odor: Odorless Melting Point: 300° MDL No.: MFCD00134536

Formula: C₈H₇NO₄
MW: 181.15 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00134536
CAS Number: 10312-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR