Betaine, anhydrous 98%
Supplier: Thermo Scientific Chemicals
B24397-0E
B24397-22
B24397-36
CAAAB24397-0EEA
741.02
CAD
CAAAB24397-0E
CAAAB24397-22
CAAAB24397-36
Betaine, anhydrous 98%
Betaine
Purity: 98%
CAS No.: 107-43-7
Molecular Formula: (CH3)3NCH2CO2
Formula Weight: 117.15
Physical Form: Cryst powder
Sensitivity: Hygroscopic
Appearance: White
Melting Point:
Formula:
C₅H₁₁NO₂ Melting Pt: >300 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012123 CAS Number: 107-43-7 EINECS: 203-490-6 Merck Index: 14,1179 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...