m-Cresol purple

Supplier: Thermo Scientific Chemicals

B24338-06 B24338-14
CAAAB24338-06EA 89.43 CAD
CAAAB24338-06 CAAAB24338-14
m-Cresol purple
m-Cresol purple

Transition interval (acid): pH 1.2 (red) to pH 2.8 (yellow); (alkaline) pH 7.4 (yellow) to pH 9.0 (purple)


CAS No.: 2303-01-7 Molecular Formula: C21H18O5S Formula Weight: 382.44 Beilstein Reference: 350314 Merck Reference: 14,2577 MDL No.: MFCD00005871

Formula: C₂₁H₁₈O₅S
Storage Temperature: Ambient
MDL Number: MFCD00005871
CAS Number: 2303-01-7
EINECS: 218-960-6
Merck Index: 14,2577

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR