Dipropyl phthalate 98%

Supplier: Thermo Scientific Chemicals
Warning

B24056-18 B24056-30
CAAAB24056-18EA 195.92 CAD
CAAAB24056-18 CAAAB24056-30
Dipropyl phthalate 98%
Dipropyl phthalate

Purity: 98% CAS No.: 131-16-8 Molecular Formula: C14H18O4 Formula Weight: 250.30 Flash Point: 110°(230°F) Refractive Index: 1.4970 Boiling Point: 168-170°/3mm Beilstein Reference: 2332522 UN No.: 3082 MDL No.: MFCD00009371

Formula: C₁₄H₁₈O₄
MW: 250.29 g/mol
Boiling Pt: 168…170 °C/3 mm
Density: 1.082
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009371
CAS Number: 131-16-8
EINECS: 205-015-8
UN: 3082
ADR: "9",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR