Dichloro(methyl)(phenyl)silane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B23999-14
B23999-36
CAAAAB23999-14EA
43.92
CAD
CAAAAB23999-14
CAAAB23999-36
Dichloro(methyl)(phenyl)silane 98%
Dichloro(methyl)(phenyl)silane
Purity: 98%
CAS No.: 149-74-6
Molecular Formula: C7H8Cl2Si
Formula Weight: 191.13
Flash Point: 82°(179°F)
Refractive Index: 1.5190
Odor: Acrid
Boiling Point: 205-206°
Beilstein Reference: 970975
UN No.: 2437
MDL No.: MFCD00000490
Formula:
C₇H₈Cl₂Si Boiling Pt: 205…206 °C Density: 1.173 Flash Pt: 82 °C(179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000490 CAS Number: 149-74-6 EINECS: 205-746-2 UN: 2437 ADR: "8",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...