Furfurylamine 99%

Supplier: Thermo Scientific Chemicals
Danger

B23975-22 B23975-36
CAAAB23975-22EA 57.35 CAD
CAAAB23975-22 CAAAB23975-36
Furfurylamine 99%
Furfurylamine

Purity: 99% CAS No.: 617-89-0 Molecular Formula: C5H7NO Formula Weight: 97.12 Flash Point: 37°(99°F) Refractive Index: 1.4900 Odor: Amine-like pH: 11.6 Melting Point: -70° Boiling Point: 145-146° Beilstein Reference: 1614 UN No.: 2526 MDL No.: MFCD00003258

Formula: C₅H₇NO
Boiling Pt: 145…146 °C
Melting Pt: –70 °C
Density: 1.050
Flash Pt: 37 °C(99 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003258
CAS Number: 617-89-0
EINECS: 210-536-9
UN: 2526
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR