6-Methoxy-2-naphthaldehyde 99%
Supplier: Thermo Scientific Chemicals
B23943-06
B23943-14
B23943-22
CAAAB23943-06EA
74.1
CAD
CAAAB23943-06
CAAAB23943-14
CAAAB23943-22
6-Methoxy-2-naphthaldehyde 99%
6-Methoxy-2-naphthaldehyde
Purity: 99%
CAS No.: 3453-33-6
Molecular Formula: C12H10O2
Formula Weight: 186.21
Melting Point: 80-82°
Beilstein Reference: 1945989
Formula:
C₁₂H₁₀O₂ Melting Pt: 80…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081152 CAS Number: 3453-33-6 EINECS: 222-377-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...