Pyridine-2-aldoxime methochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyridine-2-carboxaldoxime methochloride, Pralidoxime Chloride, 2-PAM chloride, 2-((Hydroxyimino)methyl)-1-methylpyridin-1-ium chloride

B23890-06 B23890-14
CAAAB23890-06EA 65.57 CAD
CAAAB23890-06 CAAAB23890-14 CAAAB23890-22
Pyridine-2-aldoxime methochloride 97%
Pyridine-2-aldoxime methochloride

Purity: 97% CAS No.: 51-15-0 Molecular Formula: C7H9ClN2O Formula Weight: 172.62 Melting Point: ca 223° dec. Beilstein Reference: 4163981 Merck Reference: 14,7703 MDL No.: MFCD00011981

Formula: C₇H₉ClN₂O
MW: 172.61 g/mol
Melting Pt: ca 223 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011981
CAS Number: 51-15-0
EINECS: 200-080-9
Merck Index: 14,7703

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR