3,5-Di-tert-butyl-2-hydroxybenzaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5-Di-tert-butylsalicylaldehyde

B23834-06 B23834-14 B23834-22
CAAAB23834-06EA 98.43 CAD
CAAAB23834-06 CAAAB23834-14 CAAAB23834-22
3,5-Di-tert-butyl-2-hydroxybenzaldehyde 99%
3,5-Di-tert-butyl-2-hydroxybenzaldehyde

Purity: 99% CAS No.: 37942-07-7 Molecular Formula: C15H22O2 Formula Weight: 234.34 Physical Form: Cryst solid Sensitivity: Air Sensitive Beilstein Reference: 1877810 MDL No.: MFCD00191998

Formula: C₁₅H₂₂O₂
MW: 234.34 g/mol
Melting Pt: 59…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00191998
CAS Number: 37942-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR