Diisopropyl phosphonate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diisopropyl phosphite, Di-i-propylphosphite, Diisopropyl hydrogen phosphite

B23800-14 B23800-22
CAAAB23800-14EA 78.68 CAD
CAAAB23800-14 CAAAB23800-22
Diisopropyl phosphonate 98%
Diisopropyl phosphonate

Purity: 98% CAS No.: 1809-20-7 Molecular Formula: C6H15O3P Formula Weight: 166.16 Flash Point: 112°(234°F) Refractive Index: 1.4075 Boiling Point: 72-75°/10mm MDL No.: MFCD00117905

Formula: C₆H₁₅O₃P
MW: 166.16 g/mol
Boiling Pt: 72…75 °C/10 mm
Density: 0.990
Flash Pt: 112 °C(234 °F)
Storage Temperature: Ambient
MDL Number: MFCD00117905
CAS Number: 1809-20-7
EINECS: 217-317-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR