5-Amino-2-methylbenzothiazole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Methylbenzo[d]thiazol-5-amine, 2-Methyl-1,3-benzothiazol-5-amine
B23631-04
B23631-09
CAAAB23631-04EA
122.92
CAD
CAAAB23631-04
CAAAB23631-09
CAAAB23631-18
5-Amino-2-methylbenzothiazole 99%
5-Amino-2-methylbenzothiazole
Purity: 99%
CAS No.: 13382-43-9
Molecular Formula: C8H8N2S
Formula Weight: 164.22
MDL No.: MFCD00226291
Formula:
C₈H₈N₂S MW: 164.23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00226291 CAS Number: 13382-43-9 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...