Catechol violet
Supplier: Thermo Scientific Chemicals
B23513-06
B23513-14
CAAAAB23513-06EA
101.89
CAD
CAAAAB23513-06
CAAAAB23513-14
Catechol violet
Catechol violet
CAS No.: 115-41-3
Molecular Formula: C19H14O7S
Formula Weight: 386.38
Beilstein Reference: 353938
MDL No.: MFCD00005868
Formula:
C₁₉H₁₄O₇S Storage Temperature: Ambient |
MDL Number:
MFCD00005868 CAS Number: 115-41-3 EINECS: 204-088-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...