N-(3-Aminopropyl)morpholine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-(4-Morpholinyl)propylamine, 3-Morpholinopropan-1-amine, 3-Morpholino propylamine
B23485-18
B23485-30
CAAAB23485-18EA
75.68
CAD
CAAAB23485-18
CAAAB23485-30
N-(3-Aminopropyl)morpholine 98%
N-(3-Aminopropyl)morpholine
Purity: 98%
CAS No.: 123-00-2
Molecular Formula: C7H16N2O
Formula Weight: 144.22
Flash Point: 98°(210°F)
Refractive Index: 1.4761
Melting Point: -15°
Boiling Point: 223-225°
UN No.: 2735
MDL No.: MFCD00006184
Formula:
C₇H₁₆N₂O Boiling Pt: 223…225 °C Melting Pt: –15 °C Density: 0.987 Flash Pt: 98 °C(210 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006184 CAS Number: 123-00-2 EINECS: 204-590-2 UN: 2735 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...